The physical conditions of the device are specified as follows:
Type
Specification
Range of use
Indoor use only
Ambient operating temperature
5 °C - 35 °C
Ambient storage temperature
-20 °C - 45 °C
Ambient operating humidity
20% - 80% relative humidity (non-condensing)
Ambient storage humidity
Maximum 60% relative humidity (non-condensing)
Operating altitude
Maximum 2000 m above sea level
Pollution degree
2
Power requirements
100 – 240 V AC, ± 10 %; 50/60 Hz; max. 150 W / 255 VA
Overvoltage category
II
Emission sound pressure level
typically < 50 dB(A)
Dimensions
(height x width x depth)
19.2 x 42 x 62 cm
Weight
Approx. 18 kg