You can remove the slide-in module from the enclosure of a module for transporting or shipping purposes. The enclosure remains in the system stack. To return a defective module to the factory, install the slide-in module in the enclosure of the replacement module.