Preparations
Prepare the device for transport. See Preparing the Device for Transport.
Follow these steps
  1. Observe the notes for handling and lifting the device safely.
  2. Transport the device to the new location.
  3. Install and set up the device in the system stack. Follow the instructions on mounting the system stack in the Vanquish System Operating Manual.
  4. Set up the device:
  1. Connect the device and set up flow connections (see Installation).
  2. Prepare the device for operation (see Preparing the Device for Operation).
  1. Before starting an analysis, let the device equilibrate and be sure that it is ready for operation.