To prepare the device for transport, follow these steps:
  1. Perform a long-term shut down of the device (see Long-Term Shutdown).
  2. Turn off the device with its main power switch and disconnect the power cord.
  3. Remove all cables and flow connections to other devices.
    1. Disconnect the gas inlet and exhaust tubes from the detector. See Disconnecting the Gas Inlet and Exhaust Tubes.
    2. Disconnect the waste line from the detector. See Disconnecting the Waste Line.
    3. Remove the nebulizer. See Removing the Nebulizer.
    1. Remove the device or slide-in module from the system stack as required:
      Install the carrying handles and remove the device from the Vanquish system. Follow the instructions on dismounting the system stack in the Transporting or Shipping the System section of the Vanquish System Operating Manual.
      ‑or‑
      Remove the slide-in module from the device enclosure in the system stack (see Removing the Slide-In Module).