The power switch on the device is the main switch for power on/off control. The main power switch is turned on during initial installation of the device.
For easier handling, you can use the power button on the front left of the Vanquish system base (system power button) for power on/off.
Observe the following:
  • All modules in the Vanquish system that are connected to the system base via system interlink cables are turned on or off simultaneously when the system power button is pressed.
  • When the power is on, the system power button is pressed in. When the power is off, the system power button stands out.
  • If the main power switch on a device is off, you cannot turn on the device with the system power button.
  • To turn off a device completely, you have to turn it off with the main power switch on the device. Pressing the system power button will not be sufficient to turn off the power to the device completely.
Upon power up, the detector performs a self-test. If the self-test is not successful, the status indicators are red and the detector is not ready for analysis. Check the Instrument Audit Trail for the related message and take appropriate remedial action.
After the self-test, a warm-up phase follows. During this phase, the detector monitors the temperature of the internal components.