The status LED bar on the front side of the device and the STATUS LED on the inside keypad provide information about the device status.
LED Bar
The LED bar colors provide the following information:
LED Bar | Description |
---|---|
Off (dark) | The power to the device is turned off. |
Dimmed | The doors of the device are open. |
Yellow, flashing slowly | The power to the device is turned on, but the device is not connected in the chromatography data system. |
Yellow | The LED bar is yellow when both conditions apply:
|
Green, flashing | The detector prepares for operation. This comprises the following phases:
|
Green | The LED bar is green when both conditions apply:
|
Blue, running | A data acquisition is running. |
Blue | The LED bar is blue when both conditions apply:
|
Red | A problem or error has occurred. For the related message, check the Instrument Audit Trail. For remedial action, see Troubleshooting. |
STATUS LED
The STATUS LED on the keypad inside the device provides the following information:
STATUS LED | Description |
---|---|
Off (dark) | The power to the device is turned off. |
Green | The device is functioning properly. |
Red | A problem or error has occurred. For the related message, check the Instrument Audit Trail. For remedial action, see Troubleshooting. |
For information about the LED that is present next to the GAS button on the keypad, see Keypad.