The keypad inside the device allows you to perform certain functions directly from the device. When you press a button, a short beep confirms that the function is performed.
When the device is connected in the chromatography data system, some functions may not be available from the keypad (see further down in this section).
STATUS
The STATUS LED provides a quick visual check of the operational status of the device.
When the doors are closed, the LED bar on the front side indicates the operational status.
For status details, see Status Indicators.
MUTE ALARM
Beeping alerts you when the device detects a problem, for example, leakage. To turn off the beep for the current alarm, press this button. Eliminate the source for the alarm within 10 minutes. Otherwise, beeping starts again. If the device detects a different problem, beeping alerts you again immediately.
GAS
Pushing the GAS button will turn the detector gas flow on and off.
The LED next to the button indicates the gas status:
LED | Description |
---|---|
Off (dark) | The detector gas flow and drain pump are turned off. |
Green | The detector gas flow and drain pump are turned on. |
When the Device is Connected in the Chromatography Data System
The button functionality is as follows when the device is connected in the chromatography data system:
No injection or sequence or baseline monitoring is running:
All functions are available from the keypad.
An injection or sequence or baseline monitoring is running:
The GAS button is disabled. However, the MUTE ALARM function remains available from the keypad, allowing you to turn off the beep for the current alarm.