TIP
To turn on the power to the device, follow these steps:
Check that the power button on the front left of the Vanquish system base (system power button) is pressed in. If the power button stands out, press the power button to turn on the power on the system base.
Turn on the device with its main power switch.
Turn off the device with the main power switch, when instructed to do so, for example, during maintenance. Pressing the system power button will not be sufficient to turn off the power to the device completely.
NOTICE
Make sure that the pump flow to the detector is turned off. To properly turn on the detector gas flow and only then turn on the pump flow to the detector, follow the instructions in Preparing the Device for Operation.