Leaking liquids of the device are collected in the leak tray, where they flow off through the chase on the right side of the leak tray to the drain system.
For information about how the liquid is discharged to waste through the Vanquish drain system, refer to the Vanquish System Operating Manual.