The table lists the safety symbols that appear on the device or on labels affixed to the device. Follow the safety notices in this manual to prevent the risk of operator injury or damage to the device.
Symbol | Description |
---|---|
| Indicates a potential hazard. Refer to this manual to avoid the risk of personal injury and/or to prevent damage to the device. |
— Ο | Power supply is on Power supply is off |
| Indicates alternating current. |