The following table describes the commands in the Workbook Editor.
Command | Description |
|---|---|
Update | Refreshes and updates the Results table after you edit the parameters. |
Save | Saves the target peptide workbook to the Workbook Manager. |
Delete | Deletes the components selected in the Results table from the target peptide workbook. |
Export | Exports the currently open target peptide workbook as a |
Close | Closes the Workbook Editor. |