On the Process and Review page, you can view various types of chromatograms for the peaks and/or proteoforms detected in your Top Down Analysis experiment. Each chromatogram plot displays the relative signal intensity versus retention time for the selected component.
Procedure
- Open the results of your experiment from the Queue page or from the Load Results page.
- The Process and Review page opens by default and displays the results of your Top Down Analysis experiment, including the chromatograms.
- Depending on the parameters specified in the processing method, the Chromatogram pane displays one of the following chromatograms:
- •Extracted ion chromatogram (XIC): Appears when the Calculate XIC checkbox is selected in the processing parameters (for Xtract deconvolution of full-scan MS data)
- •Total ion chromatogram (TIC): Available for all experiments.
- •Base peak chromatogram (BPC): Available for all experiments.
- See Chromatogram plot types.
- To change the chromatogram type displayed in the Chromatogram pane, right-click anywhere in the pane to open the shortcut menu, select Chromatogram Trace Type, and select the checkbox for the chromatogram type you want to display.
- See Chromatogram pane commands.
- The chromatogram mode determines the function to apply when you drag the cursor over a retention time area of the Chromatogram pane. Select the chromatogram mode in the upper right corner of the Chromatogram pane:
- Averaging: Averages the spectra for all the scans in the retention time range that you drag the cursor over (left to right) and displays them in the Source Spectrum pane.
- See View the source spectra.
- Auto Zooming: Enlarges the area (intensity and time) that you drag the cursor over without changing the view displayed in the Source Spectrum pane.
- Do one of the following:
- In the Intact Fragmentation Results table, select the row of a peak (or lower-level proteoform).
- In the Intact Deconvolution Results table, select a peak from the Peak Selection list in the title bar.
- The Chromatogram pane shows one plot (for a single loaded raw data file in the experiment) or multiple stacked plots (for multiple loaded raw data files).
- The chromatogram displays a red box around the retention time (RT) range specified for the selected (or upper level) peak.
- (Optional) Do one of the following:
- In the Intact Fragmentation Results table, expand (+/− icon) a peak row in the table and then select the row for a single proteoform.
- In the Intact Deconvolution Results table, expand (+/− icon) a component row in the table and then select the row for a single raw data file.
- The Chromatogram pane shows the chromatogram plot for the selected raw data file.