Right-click the Intact Fragmentation Results table on the Process and Review page to open a shortcut menu with the commands listed in the following table.
Command | Description |
---|---|
Export All | Exports all data in the Intact Fragmentation Results table as an Excel workbook (.xlsx). |
All Levels | Exports the results at all levels in the table. |
Proteoform Level Only | Exports only the results at the top (proteoform) level. |
Right-click the Intact Deconvolution Results table on the Process and Review page to open a shortcut menu with the commands listed in the following table.
Command | Description |
---|---|
Set As Reference Component | Sets the mass of the selected component in the Intact Deconvolution Results table as the reference mass. |
Export All | Exports all data in the Intact Deconvolution Results table as an Excel workbook (.xlsx). |
Export Checked | Exports only the selected data in the Intact Deconvolution Results table as an Excel workbook (.xlsx). |
Export Json | Exports all results in the Intact Deconvolution Results table to a text-based JavaScript file for data interchange. |
Peak and Component Levels Only | Exports only the peak information and the component-level results in the table. The exported data does not include the results at the raw data file and charge state levels. |
All Levels | Exports the peak information and results at all levels in the table. |