The BioPharma Finder application provides in-depth characterization of biotherapeutics including both proteins and oligonucleotides. With BioPharma Finder, you can perform peptide mapping, oligonucleotide, intact mass, and top-down protein analyses as well as multi-attribute method (MAM) workflows.
The following workflows are available in BioPharma Finder 5.4:
- Peptide Mapping Analysis
- Oligonucleotide Analysis
- Intact Mass Analysis
- Top Down Analysis
BioPharma Finder facilitates comprehensive interpretation and data visualization, letting you characterize your biologics with speed and ease. From novel deconvolution algorithms capable of delivering complete results to easy-to-understand data visualization tools, BioPharma Finder helps you choose the right path for protein and oligonucleotide characterization.
The BioPharma Finder software application consolidates the Thermo Scientific PepFinder™ and Thermo Scientific Protein Deconvolution™ software applications into one platform. It also includes various features from the Thermo Scientific Pinpoint™ software and ProSight Lite, a free tool developed by the Proteomics Center of Excellence at Northwestern University.
Each BioPharma Finder user guide describes a specific customizable workflow in detail and explains how you can use BioPharma Finder to interpret liquid chromatography-mass spectrometry (LC/MS) experimental results and characterize biotherapeutics.