On the page, you can import, export, and delete workbooks using the Workbook Manager.
TIP
You can access the Workbook Manager by selecting the tab on the navigation bar within the Top Down Analysis workflow.
The following table describes how to perform common Workbook Manager tasks.
Table Workbook Manager tasks
Task
Actions
Result
Edit a workbook
In the Workbook Manager table, select the checkbox for a workbook row and then select Edit on the command bar.
The Workbook Editor page opens.
Import a workbook
Select Import on the command bar. Then, in the Open dialog box, locate and select the workbook files (.wbpf) you want to import and select Open.
A confirmation message appears. Select OK to continue.
The imported workbooks are added to the Workbook Manager table.
If an imported workbook has the same name as an existing workbook in the table, the application appends "_imported" to the name.
Export a workbook
In the Workbook Manager table, select the checkbox for one or more workbook rows and then select Export on the command bar. Then, in the Save As dialog box, browse to the folder location where you want to save the workbook files (.wbpf) and select Save.
A confirmation message appears. Select OK to continue.
The selected workbooks are saved in the specified file location.
Delete a workbook
On the Workbook Manager table, select one or more workbook rows (not checkboxes) and then either select Delete on the command bar or press the DELETE key.
A confirmation message appears. Select OK to continue.
The selected workbooks are removed from the Workbook Manager table and permanently deleted from the application.