You can export the data in the ProSightBP Output Results table as an Excel worksheet (.xlsx).
Prerequisites
- You have loaded the experiment results and are viewing the ProSightBP Output Results pane on the Process and Review page.
Procedure
- On the Process and Review page, right-click anywhere in the ProSightBP Output Results table and select one of the following options on the shortcut menu:
- Export All to export all of the results in the table as an Excel worksheet (.xlsx).
- Export Checked to export only the selected results as an Excel worksheet (.xlsx).
- To select specific results, select the checkbox for a fragment ion row in the ProSightBP Output Results table.
- To select or deselect all of the rows, select or clear the checkbox in the table header row.
- Then, select one of the following sub-menu commands:
- Component Level Only to export the results only at the fragment ion (top) level in the table.
- When you select "Component Level Only", the exported results do not include the sub-component level data for a fragment ion row.
- Component and Charge State Levels to export the results at all levels in the table.
- The Save As dialog box opens.
- Browse to the file location where you want to save your results and type a file name.
- Select Save.
- The application saves the results as an Excel worksheet (.xlsx).
- The exported data reflects the current filtering or sorting of the table in the BioPharma Finder application.