Right-clicking the ProSightBP Fragment Map pane on the Process and Review page opens a shortcut menu that contains the commands listed in the following table.
Command | Description |
|---|---|
Save As (.png) | Saves the fragment map as an image file with a .png extension. |
Copy | Copies the image in the pane to the clipboard. |