Right-clicking the Intact Fragmentation Source Spectrum or Intact Deconvolution Source Spectrum pane on the Process and Review page opens a shortcut menu that contains the commands listed in the following table.
Table Intact Fragmentation Source Spectrum or Intact Deconvolution Source Spectrum pane shortcut menu
Command
Description
Reset Scale
Restores the original scale to the pane.
Copy as Displayed
Copies the image in the pane to the clipboard as currently displayed.
Copy per Global Settings
Copies the image in the pane to the clipboard based on the global image dimensions set in the Top Down Analysis Settings dialog box.
Zoom Out
Shrinks the view in the pane by a factor of 2.
Zoom In
Enlarges the view in the pane by a factor of 2.