When the BioPharma Finder application completes the processing of your experiment, you can open the results of your analysis from the Queue page or from the Load Results page.
The Process and Review page opens by default, displaying the Intact Fragmentation and Intact Deconvolution Results tables, chromatograms, deconvoluted and source spectra, ProSightBP results, and matched sequence information.