Create a new Top Down Analysis experiment on the Top Down Analysis page. On this page, enter the experiment name, load the raw data files, select a protein sequence, and select a processing method.
IMPORTANT
For Top Down Analysis experiments, only sequences assigned the category “Top Down” appear in the table.
If your protein sequence does not appear in the table, you can change its assigned category to “Top Down” using the Protein Sequence Editor.
Figure Top Down Analysis page
123456
No.DescriptionNo.Description
1
Enter experiment name
4
Select protein sequence(s)
2
Load raw data files and assign conditions
5
Select processing method
3
Specify results format
6
Open the processing method editing wizard to specify parameters and submit experiment for processing
Prerequisites
  1. You have added a protein sequence to the Sequence Manager.
Procedure
  1. On the BioPharma Finder Home page, in the Experiment Types pane, select the Top Down Analysis workflow link.
  2. The Top Down Analysis page opens.
  3. In the Top Down Analysis Definition area, type the name of the experiment in the Experiment Name box.
  4. TIP
    Experiment names can contain up to 50 characters, including alphanumeric, space, underscore (“_”), and period (“.”) characters.
  5. IMPORTANT
    You can overwrite a completed experiment by entering the same experiment name.
    You cannot overwrite an experiment that is currently submitted to the run queue or is actively processing or canceling in the run queue.
  6. In the Load Raw Data area, select the Select Raw Data (“...”) button.
  7. The Add Analysis File(s) dialog box opens.
  8. NOTE
    The default file location for raw data files is C:\Xcalibur\data.
  9. In the Add Analysis File(s) dialog box, locate and select the raw data files (.raw) you want to add to the experiment.
  10. TIP
    To add a set of raw data files to the experiment (for example, replicates of the same sample or samples with various experiment conditions), select contiguous files using the SHIFT key or noncontiguous files using the CTRL key.
  11. Select Open.
  12. The application adds the imported raw data files to the table in the Load Raw Data area.
  13. The order of the files in this table defines the order that they appear in the Results tables on the Process and Review page.
  14. In the Load Raw Data area, in the Condition box, type the experiment condition for each raw data file, separated by a space.
  15. If an entered condition matches a portion of a raw data file name, the application automatically assigns that condition to one or more matching raw data files.
  16. To change the condition assigned to a raw data file, use the drop-down menu in the Condition column next to the corresponding raw file name to select a different condition.
  17. In the Reference Condition box, select the reference condition for the experiment from the drop-down menu.
  18. NOTE
    The default reference condition is the first entered condition in the Condition box.
  19. The application uses the indicated reference condition to calculate various analytical ratios during processing. These ratios are reported in the Results table(s) on the Process and Review page.
  20. If you load multiple raw data files for an experiment, in the Result Format for Multiple Raw Files area, select either the Batch Processing or Multiconsensus option.
  21. TIP
    For more information on these results formats, see Result formats: Batch Processing and Multiconsensus.
  22. NOTE
    For Intact Mass Analysis and Top Down Analysis experiments, you can process up to 10 raw data files using the Multiconsensus results format.
  23. In the Sequence area, select the checkbox in the table row for up to 10 sequences you want to assign to the experiment.
  24. The application applies the selected sequence to the experiment during processing.
  25. In the Processing Method area, select the checkbox for the processing method you want to use or customize for your experiment.
  26. Then, select the Start Processing button to proceed.
  27. The processing method editing wizard opens and prompts you to edit the method parameters and save them to a new file to create a custom method for your experiment.