The following table lists the one-letter codes for amino acids used by the BioPharma Finder application.
NOTE
The one-letter amino acid codes are not case-sensitive.
Table Amino acid letter codes
One-letter code
Amino acid
A
alanine
R
arginine
N
asparagine
D
aspartic acid
C
cysteine
E
glutamic acid
Q
glutamine
G
glycine
H
histidine
I
isoleucine
L
leucine
K
lysine
M
methionine
F
phenylalanine
P
proline
S
serine
T
threonine
W
tryptophan
Y
tyrosine
V
valine