The following table lists the N-linked glycans that the BioPharma Finder application includes in the Define Modification List window.
Table N-Linked glycans (side chain modifications) in the Define Modification List window
#
Glycan name
Formula
Average mass
Monoisotopic mass
1
A1G0
C42H69N3O30
1095.40
1095.40
2
A1G0F
C48H79N3O34
1241.45
1241.45
3
A1G0M4
C48H79N3O35
1257.45
1257.45
4
A1G0M4F
C54H89N3O39
1403.51
1403.51
5
A1G0M5
C54H89N3O40
1419.50
1419.50
6
A1G0M5F
C60H99N3O44
1565.56
1565.56
7
A1G1
C48H79N3O35
1257.45
1257.45
8
A1G1F
C54H89N3O39
1403.51
1403.51
9
A1G1M4
C54H89N3O40
1419.50
1419.50
10
A1G1M4F
C60H99N3O44
1565.56
1565.56
11
A1G1M5
C60H99N3O45
1581.56
1581.56
12
A1G1M5F
C66H109N3O49
1727.61
1727.61
13
A1S1
C59H96N4O43
1548.54
1548.54
14
A1S1F
C65H106N4O47
1694.60
1694.60
15
A1S1M4
C65H106N4O48
1710.60
1710.60
16
A1S1M4F
C71H116N4O52
1856.66
1856.66
17
A1S1M5
C71H116N4O53
1872.65
1872.65
18
A1S1M5F
C77H126N4O57
2018.71
2018.71
19
A1Sg1
C59H96N4O44
1564.54
1564.54
20
A1Sg1F
C65H106N4O48
1710.60
1710.60
21
A2G0
C50H82N4O35
1298.48
1298.48
22
A2G0B
C58H95N5O40
1501.56
1501.56
23
A2G0F
C56H92N4O39
1444.53
1444.53
24
A2G0FB
C64H105N5O44
1647.61
1647.61
25
A2G0M4
C56H92N4O40
1460.53
1460.53
26
A2G0M5
C62H102N4O45
1622.58
1622.58
27
A2G0M5F
C68H112N4O49
1768.64
1768.64
28
A2G1
C56H92N4O40
1460.53
1460.53
29
A2G1B
C64H105N5O45
1663.61
1663.61
30
A2G1F
C62H102N4O44
1606.59
1606.59
31
A2G1FB
C70H115N5O49
1809.67
1809.67
32
A2G1M4
C62H102N4O45
1622.58
1622.58
33
A2G1M4F
C68H112N4O49
1768.64
1768.64
34
A2G1M5
C68H112N4O50
1784.63
1784.63
35
A2G1M5F
C74H122N4O54
1930.69
1930.69
36
A2G2
C62H102N4O45
1622.58
1622.58
37
A2G2B
C70H115N5O50
1825.66
1825.66
38
A2G2F
C68H112N4O49
1768.64
1768.64
39
A2G2FB
C76H125N5O54
1971.72
1971.72
40
A2G2M4
C68H112N4O50
1784.63
1784.63
41
A2G2M4F
C74H122N4O54
1930.69
1930.69
42
A2G2M5
C74H122N4O55
1946.69
1946.69
43
A2G2M5F
C80H132N4O59
2092.75
2092.75
44
A2S1G0
C67H109N5O48
1751.62
1751.62
45
A2S1G0B
C75H122N6O53
1954.70
1954.70
46
A2S1G0F
C73H119N5O52
1897.68
1897.68
47
A2S1G0FB
C81H132N6O57
2100.76
2100.76
48
A2S1G0M4
C73H119N5O53
1913.68
1913.68
49
A2S1G0M4F
C79H129N5O57
2059.73
2059.73
50
A2S1G0M5
C79H129N5O58
2075.73
2075.73
51
A2S1G0M5F
C85H139N5O62
2221.79
2221.79
52
A2S1G1
C73H119N5O53
1913.68
1913.68
53
A2S1G1B
C81H132N6O58
2116.76
2116.76
54
A2S1G1F
C79H129N5O57
2059.73
2059.73
55
A2S1G1FB
C87H142N6O62
2262.81
2262.81
56
A2S1G1M4
C79H129N5O58
2075.73
2075.73
57
A2S1G1M4F
C85H139N5O62
2221.79
2221.79
58
A2S1G1M5
C85H139N5O63
2237.78
2237.78
59
A2S1G1M5F
C91H149N5O67
2383.84
2383.84
60
A2S2
C84H136N6O61
2204.77
2204.77
61
A2S2B
C92H149N7O66
2407.85
2407.85
62
A2S2F
C90H146N6O65
2350.83
2350.83
63
A2S2FB
C98H159N7O70
2553.91
2553.91
64
A2S2M4
C90H146N6O66
2366.83
2366.83
65
A2S2M4F
C96H156N6O70
2512.88
2512.88
66
A2S2M5
C96H156N6O71
2528.88
2528.88
67
A2S2M5F
C102H166N6O75
2674.94
2674.94
68
A2Sg1G0
C67H109N5O49
1767.62
1767.62
69
A2Sg1G0F
C73H119N5O53
1913.68
1913.68
70
A2Sg1G1
C73H119N5O54
1929.67
1929.67
71
A2Sg1G1F
C79H129N5O58
2075.73
2075.73
72
A2Sg1S1
C84H136N6O62
2220.77
2220.77
73
A2Sg1S1F
C90H146N6O66
2366.83
2366.83
74
A2Sg2
C84H136N6O63
2236.76
2236.76
75
A2Sg2F
C90H146N6O67
2382.82
2382.82
76
A3G0
C58H95N5O40
1501.56
1501.56
77
A3G0F
C64H105N5O44
1647.61
1647.61
78
A3G1
C64H105N5O45
1663.61
1663.61
79
A3G1F
C70H115N5O49
1809.67
1809.67
80
A3G2
C70H115N5O50
1825.66
1825.66
81
A3G2F
C76H125N5O54
1971.72
1971.72
82
A3G3
C76H125N5O55
1987.71
1987.71
83
A3G3F
C82H135N5O59
2133.77
2133.77
84
A3S1G0
C75H122N6O53
1954.70
1954.70
85
A3S1G0F
C81H132N6O57
2100.76
2100.76
86
A3S1G1
C81H132N6O58
2116.76
2116.76
87
A3S1G1F
C87H142N6O62
2262.81
2262.81
88
A3S1G2
C87H142N6O63
2278.81
2278.81
89
A3S1G2F
C93H152N6O67
2424.87
2424.87
90
A3S2G0
C92H149N7O66
2407.85
2407.85
91
A3S2G0F
C98H159N7O70
2553.91
2553.91
92
A3S2G1
C98H159N7O71
2569.90
2569.90
93
A3S2G1F
C104H169N7O75
2715.96
2715.96
94
A3S3
C109H176N8O79
2861.00
2861.00
95
A3S3F
C115H186N8O83
3007.06
3007.06
96
A3Sg1G0
C75H122N6O54
1970.70
1970.70
97
A3Sg1G0F
C81H132N6O58
2116.76
2116.76
98
A3Sg1G1
C81H132N6O59
2132.75
2132.75
99
A3Sg1G1F
C87H142N6O63
2278.81
2278.81
100
A3Sg1G2
C87H142N6O64
2294.80
2294.80
101
A3Sg1G2F
C93H152N6O68
2440.86
2440.86
102
A3Sg1S1G0
C92H149N7O67
2423.85
2423.85
103
A3Sg1S1G0F
C98H159N7O71
2569.90
2569.90
104
A3Sg1S1G1
C98H159N7O72
2585.90
2585.90
105
A3Sg1S1G1F
C104H169N7O76
2731.96
2731.96
106
A3Sg1S2
C109H176N8O80
2876.99
2876.99
107
A3Sg1S2F
C115H186N8O84
3023.05
3023.05
108
A3Sg2G0
C92H149N7O68
2439.84
2439.84
109
A3Sg2G0F
C98H159N7O72
2585.90
2585.90
110
A3Sg2G1
C98H159N7O73
2601.89
2601.89
111
A3Sg2G1F
C104H169N7O77
2747.95
2747.95
112
A3Sg2S1
C109H176N8O81
2892.99
2892.99
113
A3Sg2S1F
C115H186N8O85
3039.05
3039.05
114
A3Sg3
C109H176N8O82
2908.98
2908.98
115
A3Sg3F
C115H186N8O86
3055.04
3055.04
116
A4G0
C66H108N6O45
1704.63
1704.63
117
A4G0F
C72H118N6O49
1850.69
1850.69
118
A4G1
C72H118N6O50
1866.69
1866.69
119
A4G1F
C78H128N6O54
2012.75
2012.75
120
A4G2
C78H128N6O55
2028.74
2028.74
121
A4G2F
C84H138N6O59
2174.80
2174.80
122
A4G3
C84H138N6O60
2190.79
2190.79
123
A4G3F
C90H148N6O64
2336.85
2336.85
124
A4G4
C90H148N6O65
2352.85
2352.85
125
A4G4F
C96H158N6O69
2498.90
2498.90
126
A4S1G0
C83H135N7O58
2157.78
2157.78
127
A4S1G0F
C89H145N7O62
2303.84
2303.84
128
A4S1G1
C89H145N7O63
2319.84
2319.84
129
A4S1G1F
C95H155N7O67
2465.89
2465.89
130
A4S1G2
C95H155N7O68
2481.89
2481.89
131
A4S1G2F
C101H165N7O72
2627.95
2627.95
132
A4S1G3
C101H165N7O73
2643.94
2643.94
133
A4S1G3F
C107H175N7O77
2790.00
2790.00
134
A4S2G0
C100H162N8O71
2610.93
2610.93
135
A4S2G0F
C106H172N8O75
2756.99
2756.99
136
A4S2G1
C106H172N8O76
2772.98
2772.98
137
A4S2G1F
C112H182N8O80
2919.04
2919.04
138
A4S2G2
C112H182N8O81
2935.04
2935.04
139
A4S2G2F
C118H192N8O85
3081.09
3081.09
140
A4S3G0
C117H189N9O84
3064.08
3064.08
141
A4S3G0F
C123H199N9O88
3210.14
3210.14
142
A4S3G1
C123H199N9O89
3226.13
3226.13
143
A4S3G1F
C129H209N9O93
3372.19
3372.19
144
A4S4
C134H216N10O97
3517.23
3517.23
145
A4S4F
C140H226N10O101
3663.29
3663.29
146
A4Sg1G0
C83H135N7O59
2173.78
2173.78
147
A4Sg1G0F
C89H145N7O63
2319.84
2319.84
148
A4Sg1G1
C89H145N7O64
2335.83
2335.83
149
A4Sg1G1F
C95H155N7O68
2481.89
2481.89
150
A4Sg1G2
C95H155N7O69
2497.88
2497.88
151
A4Sg1G2F
C101H165N7O73
2643.94
2643.94
152
A4Sg1G3
C101H165N7O74
2659.94
2659.94
153
A4Sg1G3F
C107H175N7O78
2805.99
2805.99
154
A4Sg1S1G0
C100H162N8O72
2626.93
2626.93
155
A4Sg1S1G0F
C106H172N8O76
2772.98
2772.98
156
A4Sg1S1G1
C106H172N8O77
2788.98
2788.98
157
A4Sg1S1G1F
C112H182N8O81
2935.04
2935.04
158
A4Sg1S1G2
C112H182N8O82
2951.03
2951.03
159
A4Sg1S1G2F
C118H192N8O86
3097.09
3097.09
160
A4Sg1S2G0
C117H189N9O85
3080.07
3080.07
161
A4Sg1S2G0F
C123H199N9O89
3226.13
3226.13
162
A4Sg1S2G1
C123H199N9O90
3242.13
3242.13
163
A4Sg1S2G1F
C129H209N9O94
3388.19
3388.19
164
A4Sg1S3
C134H216N10O98
3533.22
3533.22
165
A4Sg1S3F
C140H226N10O102
3679.28
3679.28
166
A4Sg2G0
C100H162N8O73
2642.92
2642.92
167
A4Sg2G0F
C106H172N8O77
2788.98
2788.98
168
A4Sg2G1
C106H172N8O78
2804.97
2804.97
169
A4Sg2G1F
C112H182N8O82
2951.03
2951.03
170
A4Sg2G2
C112H182N8O83
2967.03
2967.03
171
A4Sg2G2F
C118H192N8O87
3113.08
3113.08
172
A4Sg2S1G0
C117H189N9O86
3096.07
3096.07
173
A4Sg2S1G0F
C123H199N9O90
3242.13
3242.13
174
A4Sg2S1G1
C123H199N9O91
3258.12
3258.12
175
A4Sg2S1G1F
C129H209N9O95
3404.18
3404.18
176
A4Sg2S2
C134H216N10O99
3549.22
3549.22
177
A4Sg2S2F
C140H226N10O103
3695.28
3695.28
178
A4Sg3G0
C117H189N9O87
3112.06
3112.06
179
A4Sg3G0F
C123H199N9O91
3258.12
3258.12
180
A4Sg3G1
C123H199N9O92
3274.12
3274.12
181
A4Sg3G1F
C129H209N9O96
3420.18
3420.18
182
A4Sg3S1
C134H216N10O100
3565.21
3565.21
183
A4Sg3S1F
C140H226N10O104
3711.27
3711.27
184
A4Sg4
C134H216N10O101
3581.21
3581.21
185
A4Sg4F
C140H226N10O105
3727.27
3727.27
186
Gn
C8H13N1O5
203.08
203.079
187
GnF
C14H23N1O9
349.14
349.137
188
M3
C34H56N2O25
892.32
892.317
189
M4
C40H66N2O30
1054.37
1054.37
190
M5
C46H76N2O35
1216.42
1216.42
191
M6
C52H86N2O40
1378.48
1378.48
192
M7
C58H96N2O45
1540.53
1540.53
193
M8
C64H106N2O50
1702.58
1702.58
194
M9
C70H116N2O55
1864.63
1864.63
195
G0_G0F
C106H174N8O74S0
2744.54
2743.010
196
G0F_G0F
C112H184N8O78S0
2890.68
2889.068
197
G0F_G1F
C118H194N8O83S0
3052.82
3051.121
198
G1F_G1F
C124H204N8O88S0
3214.96
3213.173
199
G1F_G2F
C130H214N8O93S0
3377.10
3375.226
200
G2F_G2F
C136H224N8O98S0
3539.24
3537.279