NOTE
Custom filter files created in BioPharma Finder version 5.1 or earlier can not be applied to results generated in BioPharma Finder 5.4.
You must recreate custom filter files in BioPharma Finder 5.4 to apply them to results generated in BioPharma Finder 5.4.
Procedure
  1. In the Results table on the Process and Review page, right-click anywhere in the table and select Filters > Apply from the shortcut menu.
  2. The Open dialog box opens.
  3. Browse to the folder location on your computer where you saved the custom filter file.
  4. Custom filter files contain the ".cfg" extension.
  5. Select Open.
  6. The application applies all of the filters saved in the file to the Results table.