The following table lists the various filter operators for the different types of data in the table columns.
Table Filter operators
Type of data
Filter operator
Condition description
Textual and Numerical
Equals
Data is equal to the selected filter operand or a value that you enter in the operand box.
Not equals
Data is not equal to the selected filter operand or a value that you enter in the operand box.
Less than
Data is less than the selected filter operand or a value that you enter in the operand box.
Less than or equal to
Data is less than or equal to the selected filter operand or a value that you enter in the operand box.
Greater than
Data is greater than the selected filter operand or a value that you enter in the operand box.
Greater than or equal to
Data is greater than the selected filter operand or a value that you enter in the operand box.
In
Data contains the selected filter operand or a substring that you enter in the operand box.
Not in
Data does not contain the selected filter operand or a substring that you enter in the operand box.
Textual
Contains
Data contains the selected filter operand or text that you enter in the operand box.
Does not contain
Data does not contain the selected filter operand or text that you enter in the operand box.
Like (wildcards)
Data is like the selected filter operand or text and a wildcard character that you enter in the operand box. Use these wildcard characters:
  • "*" to replace any characters
  • "?" to replace one character
Not like (wildcards)
Data are not like the selected filter operand or text and a wildcard character that you enter in the operand box. Use these wildcard characters:
  • "*" to replace any characters
  • "?" to replace one character
Match (regular expression)
Data matches a regular expression that you enter in the operand box.
Does not match (regular expression)
Data does not match a regular expression that you enter in the operand box.
Starts with
Data starts with the selected filter operand or text that you enter in the operand box.
Does not start with
Data does not start with the selected filter operand or text that you enter in the operand box.
Ends with
Data ends with the selected filter operand or text that you enter in the operand box.
Does not end with
Data does not end with the selected filter operand or text that you enter in the operand box.
Numerical
Top
Enter a number, n, in the operand box.
The filter column displays only the highest n numbers out of all the numbers in that column.
Bottom
Enter a number, m, in the operand box.
The filter column displays only the lowest m numbers out of all the numbers in that column.
Top Percentile
Enter a number, x, in the operand box.
The filter column displays only the highest x% out of all the numbers in that column.
Bottom Percentile
Enter a number, y, in the operand box.
The filter column displays only the lowest y% out of all the numbers in that column.