You can filter data rows in the Results table on the Process and Review page to clearly visualize specific results.
Prerequisites
- You have processed your experiment and are viewing the Results table on the Process and Review page.
Procedure
- In the Results table, identify the column you want to filter.
- In the filter row (below the column headers), select a filter operator from the drop-down list.
Figure Select a filter operator (text-based column) Figure Select a filter operator (numerical column) - See Table filter operators.
- Select a filter operand from the list of options for a particular filter column (if available), or type a value in the operand box for a condition.
Figure Select a filter operand - See Table filter operands.
- The table displays only the rows with values that fulfill the selected condition in the filter column.
- Some of the operators do not apply to all of the available operands, or they can have other special operands.
- If you select operators and operands for multiple columns, the table shows only the rows with values that fulfill all of the conditions in the selected filter columns.