Procedure
- On the BioPharma Finder Home page, in the Experiment Types pane, select the appropriate workflow link.
- The corresponding workflow page opens.
- Select Import Method on the command bar.
- The Import Method dialog box opens.
Figure Import Method button on the Intact Mass Analysis page - In the Import Method dialog box, locate and select the processing method files (.
methodbpf) you want to import. TIP
For clarity, file names are appended according to their respective workflows.
TIP
To import multiple files simultaneously, select contiguous files using the SHIFT key or noncontiguous files using the CTRL key.
- Select Open.
- A confirmation message appears.
- The application displays the imported processing methods in the Processing Method table.
- If an imported method has the same name as an existing method in the table, the applications appends "
_imported" to the name.