When you run the BioPharma Finder application, temporary processing files are saved to your local computer. If these files cause disk space issues, you can manually trigger data space optimization via the Ardia Platform menu. Alternatively, the application will automatically perform a forced purge if the available disk space reaches a critical threshold. In both instances, temporary processing files are prioritized and then deleted to free up disk space.
IMPORTANT
The BioPharma Finder application does not display warning message before starting a forced purge.
A forced purge proceeds in the background and deletes files from the local cache until the appropriate disk space is available. The application prioritizes files during a forced purge based on file size, usage count (how often is the file being used), reference count (how many experiments use the file), and when the file was last used.
The BioPharma Finder application provides warning messages or performs a forced purge action if the disk space reaches the threshold levels described in the table below.
Table Disk space thresholds for forced purge actions in BioPharma Finder
Threshold
Description
Storage
Disk space warning
The storage limit at which the application first prompts you optimize disk space
< 25% of total disk space available
Forced start purge
The storage limit at which a forced purge starts
< 10% of total disk space available
Maximum purge limit
The storage limit at which a forced purge stop.
= 40% of total disk space available
Minimum disk space used before starting purge
The storage limit for BioPharma Finder data files at which the application performs a forced purge.
< 20% of total disk space is occupied by application files
If your computer is running low on disk space but the BioPharma Finder application is not the primary cause, the application provides a warning message and closes. You must first free up space by deleting unnecessary files from other applications or by using tune-up options that are available on your local computer. After cleanup is completed, you can start the application and perform data space optimization as needed.
Optimize disk space
Procedure
  1. On the BioPharma Finder Home page, select Ardia Platform > Data Space Optimization > Optimize Space.
  2. When prompted, select Yes to continue.
Purge all BioPharma Finder files
Procedure
  1. On the BioPharma Finder Home page, select Ardia Platform > Data Space Optimization > Clear All.
  2. When prompted, select Yes to continue.