On the Load Results page, use the Working List to organize experiments in user-created folders.
TIP
You can also open, delete, import, and export experiments from the Working List, using the same task actions described for the Master List.
The following table describes how to perform common Working List tasks.
Task | Actions | Result |
|---|---|---|
Create a new folder | Select the Home folder and then select Add. In the Add New Folder dialog box, type the folder name and then select Add. | The new folder appears in the Working List. |
Create a new subfolder | Select the folder to which you want to add a subfolder and then select Add. In the Add New Folder dialog box, type the folder name and then select Add. | The new subfolder appears within the parent folder in the Working List. |
Rename a folder | Select the folder you want to rename and then select Edit in the Experiment Manager pane. In the Edit Folder Name dialog box, type the new name and then select Edit again. | The folder is renamed. |
Delete a folder | Select the folder you want to delete and then select Delete in the Experiment Manager pane. Then, in the Delete Selected Folder dialog box, choose the appropriate sub-command:
| For the Delete folder option, the folder is deleted and any contained experiments are moved back to the Working List. For the Delete folder and results from Master List option, the folder is deleted and any contained experiments are permanently deleted from the Master List. |
Add an experiment to a folder | Select the rows of the experiments you want to add to a folder and then drag and drop the experiment results to the destination folder. Use the CTRL key to select multiple experiment rows simultaneously. | The experiment appears in the destination folder and disappears from the Working List. |
Delete an experiment | Select the checkboxes for the experiments you want to move and then select Delete on the command bar. In the Delete Selected Results from Folder dialog box, choose the appropriate sub-command:
| For the Delete results from folder sub-command, the selected experiments are removed from the current folder and returned to the Working List. For the Delete results from folder and Master List sub-command, the selected experiments are removed from the current folder and Master List and are permanently deleted from the application. |
Move an experiment from one folder to another | Select the checkboxes for the experiments you want to move and then select Delete on the command bar. In the Delete Selected Results from Folder dialog box, select the Delete results from folder option. Then, from the Working List, add the returned experiment to the desired destination folder. | The experiments are removed from one folder and then added to another. |