For a wide variety of LC/MS and UHPLC/MS-grade solvents and consumables for purchase, visit http://www.fishersci.com.
IMPORTANT
Only use LC/MS-grade or better solvents and additives.
Item | Part number |
|---|---|
Solvents | |
Isopropanol, Optima™ LC/MS, 1 L | A461-1 |
Acetonitrile, UHPLC-MS, 1 L | A9561 |
Methanol, UHPLC-MS, 1 L | A4581 |
Water, UHPLC-MS, 1 L | W81 |
Additives | |
Acetic acid, Optima™ LC/MS, 1 mL ampule | A113-1AMP |
Ammonium acetate, Optima™ LC/MS, 50 g | A11450 |
Ammonium formate, Optima™ LC/MS, 50 g | A11550 |
Formic acid, Optima™ LC/MS, 10 × 1 mL ampules | A117-10X1AMP |