The following figure shows the recommended instrument layout for the TSQ Altis Plus MS and TSQ Altis MS with data system, LC system, and oil forepumps. The other TSQ Series II instruments have the same layout, except that each has only one oil forepump.
Not all connections are shown. The connections for the syringe pump and the divert/inject valve are not included in the following figure. For installation instructions, refer to the TSQ Series II Getting Connected Guide.
Figure Workbenches for a TSQ Altis Plus MS, data system, and optional Vanquish UHPLC system
12345679810
No.
Description
No.
Description
1
Vanquish UHPLC system, 53 cm (21 in.) width
2
TSQ Altis Plus MS, 76 cm (30 in.) width
3
Data system example, 85 cm (34 in.) width
4
External vent for the forepump exhaust
5
External vent for the solvent trap (waste)
6
Electrical outlets for the LC system (120 or 230 Vac)
7
Solvent waste container
8
Forepumps
9
Separate electrical outlets for the instrument and forepumps (230 Vac circuits A, B, and C)
10
Electrical outlets for the data system (120 or 230 Vac)