Electronic equipment carriers that specialize in the handling and transport of delicate machinery ship the instrument to your site. When the instrument arrives, move it to a protected indoor location and do not open the shipping containers. A Thermo Fisher Scientific service engineer will unpack and inspect the containers during the scheduled installation visit.
NOTE
Contact Thermo Fisher Scientific Technical Support if you have questions or need assistance moving your instrument shipment.
For additional details, refer to the topics below: