API ion transfer tube
A tube assembly that assists in desolvating ions that are produced by the ESI, NSI, or APCI probe.
API ion transfer tube offset voltage
A DC voltage applied to the ion transfer tube. The voltage is positive for positive ions and negative for negative ions.
API source
The sample interface between the liquid chromatograph (LC) and the mass spectrometer (MS).
atmospheric pressure chemical ionization (APCI)
A soft ionization technique done in an ion source operating at atmospheric pressure. Electrons from a corona discharge initiate the process by ionizing the mobile phase vapor molecules. A reagent gas forms, which efficiently produces positive and negative ions of the analyte through a complex series of chemical reactions.
atmospheric pressure ionization (API)
Ionization performed at atmospheric pressure by using atmospheric pressure chemical ionization (APCI), electrospray ionization (ESI), or nanoelectrospray ionization (nanoESI or NSI).
atmospheric pressure photoionization (APPI)
A soft ionization technique in which an ion is generated from a molecule when it interacts with a photon from a light source.
auxiliary gas
The outer-coaxial gas (nitrogen) that assists the sheath (inner-coaxial) gas in dispersing and/or evaporating sample solution as the sample solution exits the APCI (optional), APPI (optional), ESI, or H-ESI spray insert. The mass spectrometer heats this gas to the user-specified vaporizer temperature.
centroid data
Data used to represent mass spectral peaks in terms of two parameters: the centroid (the weighted center of mass) and the intensity. The data is displayed as a bar graph. The normalized area of the peak provides the mass intensity data.
charge state
The imbalance between the number of protons (in the nuclei of the atoms) and the number of electrons that a molecular species (or adduct ion) possesses. If the species possesses more protons than electrons, its charge state is positive. If it possesses more electrons than protons, its charge state is negative.
collision energy
The energy used when ions collide with the collision gas.
collision gas
A neutral gas used to undergo collisions with ions.
collision-induced dissociation (CID)
A method of fragmentation where molecular ions are accelerated to high-kinetic energy and then allowed to collide with neutral gas molecules such as helium or nitrogen. The collisions break the bonds and fragment the ions into smaller pieces.
contact closure connection
The cable connection is from the external peripheral device to the mass spectrometer contact closure pins (Start In and Ground). The external device sends the contact closure (start) signal to the mass spectrometer.
conversion dynode
A highly polished metal surface that converts ions from the mass analyzer into secondary electrons, which enter the electron multiplier.
divert/inject valve
A modular valve that can be plumbed as a divert valve to control the flow direction or as a loop injector for flow injection analysis.
electron multiplier
A device used for current amplification through the secondary emission of electrons. Electron multipliers can have a discrete dynode or a continuous dynode.
electrospray ionization (ESI)
A soft ionization technique operating at atmospheric pressure. Ions are generated in solution and a high voltage is applied to generate small droplets that are then evaporated until all ions are in the gas phase.
flow rate, syringe pump status
The syringe pump injection flow rate in milliliters per minute (mL/min) or microliters per minute (μL/min) for the current sample, as defined in the current experiment method.
forevacuum pump, forepump
The pump that evacuates the foreline. A rotary-vane pump is a type of forepump. It might also be referred to as a backing, mechanical, roughing, or vacuum pump.
fragment ion
A charged dissociation product of an ionic fragmentation. Such an ion can dissociate further to form other charged molecular or atomic species of successively lower formula weights.
Full Width at Half Maximum (FWHM)
The width of a mass spectrum peak at fifty percent of the peak's height. This value determines peak resolution and is expressed as a function of mass. This value is calculated by dividing the mass-to-charge ratio (m/z) of the peak by the peak's width at fifty percent of the peak's height.
full-scan type
Provides a full mass spectrum of each analyte or parent ion. With the full-scan type, the mass analyzer is scanned from the first mass to the last mass without interruption. Also known as single-stage full-scan type.
heated-electrospray ionization (H-ESI)
A type of atmospheric pressure ionization that converts ions in solution into ions in the gas phase by using electrospray (ESI) in combination with heated auxiliary gas.
ion detection system
A high sensitivity, off-axis system for detecting ions. It produces a high signal-to-noise ratio (S/N) and allows for switching of the voltage polarity between positive ion and negative ion modes of operation. The ion detection system includes one ±12 kVdc conversion dynode and a discrete dynode electron multiplier.
ion isolation
A step in the quadrupole Q1 mass analysis where the mass analyzer ejects all ions except for the ions of interest.
ion optics
Focuses and transmits ions from the API source to the mass analyzer.
ion polarity mode
The mass spectrometer can operate in either of two ion polarity modes: positive or negative.
ion sweep cone
A removable cone-shaped metal cover that fits on top of the API ion transfer tube and acts as a physical barrier to protect the entrance of the tube.
lens
A metal disk with a circular hole in the center that allows the ion beam to pass.
mass analysis
A process that produces a mixture of ionic species that is then separated according to the mass-to-charge ratios (m/z) of the ions to produce a mass spectrum.
mass analyzer
A device that determines the mass-tocharge ratios (m/z) of ions by one of a variety of techniques.
mass analyzer DC offset voltage
A DC voltage that is applied to the mass analyzer electrodes to help draw ions in from the ion optics. This voltage defines the translational kinetic energy of the ions as they enter the mass analyzer. The automated calibration process tunes these values for positive and negative ions.
mass spectrometer
An instrument that ionizes sample molecules and then measures and analyses the ions according to their mass-to-charge ratio (m/z). The resulting mass spectrum is a characteristic pattern for the identification of a molecule.
mass spectrum
A graphical representation (plot) of measured ion abundance versus mass-to-charge ratio. The mass spectrum is a characteristic pattern for the identification of a molecule and is helpful in determining the chemical composition of a sample.
mass-to-charge ratio (m/z)
Used to denote the quantity formed by dividing the mass of an ion (in u or Da) by the number of charges carried by the ion. For example, for the ion C7H7²⁺, m/z=45.5.
molecular ion
An ion formed by the removal (positive ion) or addition (negative ion) of one or more electrons to/from a molecule without fragmentation of the molecular structure.
multipole
A symmetrical, parallel array of (usually) four, six, or eight cylindrical rods that acts as an ion transmission device. An RF voltage and a DC offset voltage are applied to the rods to create an electrostatic field that efficiently transmits ions along the axis of the multipole rods.
multipole DC offset voltage
A DC voltage applied to a multipole rod assembly. The multipole DC offset voltage helps to define the translational kinetic energy of the ions within the assembly.
multipole RF voltage
The amplitude of the RF voltage applied to the multipoles.
nanoelectrospray ionization (nano ESI or NSI)
A type of electrospray (ESI) that accommodates very low flow rates of sample and solvent on the order of 1 to 20 nL/min (for static nanospray) or 100 to 1000 nL/min (for dynamic nanoelectrospray, also called nanoESI nanoLC gradient separation).
neutral loss mass
The mass of the neutral species that is lost by the precursor ion in a neutral loss experiment.
Neutral loss scan mode
A scan mode that links together Q1 and Q3 scans so that they are scanned at the same rate over scan ranges of the same width. However, the respective mass ranges are offset by a selected mass so that the Q3 scan is a selected number of mass units lower than the Q1 scan.
peak threshold
The minimum number of intensity counts per sampling interval that is required before a signal is recorded.
peak width
The distance across a peak measured at a selected peak-height level, in minutes or mass units. The peak-height level is usually specified as a percentage of the maximum peak height.
precursor ion
An electrically charged molecular species that can dissociate to form fragments. The fragments can be electrically charged or neutral species. A precursor ion can be a molecular ion or an electrically charged fragment of a molecular ion.
precursor mass
The mass-to-charge ratio of a precursor ion. The location of the center of a target precursor-ion peak in mass-to-charge ratio (m/z) units.
product ion
An electrically charged fragment of an isolated precursor ion. In general, product ions have a direct relationship to a particular precursor ion and can correlate to a unique state of the precursor ion.
product mass
The mass-to-charge ratio of a product ion. The location of the center of a target production peak in mass-to-charge ratio (m/z) units.
profile data
Data representing mass spectral peaks as point-to-point plots, with each point having an associated intensity value.
qualitative analysis
Chemical analysis designed to determine the identity of the components of a substance.
quantitative analysis
Chemical analysis designed to determine the quantity or concentration of a specific substance in a sample.
relative standard deviation (RSD)
A measure of the dispersion of a group of measurements relative to the mean of the group. Relative standard deviation is expressed as a percentage of the average value. The percent standard deviation is calculated as: %RSD = 100 × (S / X) where S is the standard deviation and X is the sample mean.
retention time (RT)
The time after injection at which a compound elutes. The total time that the compound is retained on the chromatograph column.
RF lens
A multipole rod assembly that is operated with only radio frequency (RF) voltage on the rods. In this type of device, virtually all ions have stable trajectories and pass through the assembly.
scan
Composed of one or more microscans. Each microscan is one mass analysis (ion injection and storage/scan-out of ions) followed by ion detection. After the microscans are summed, the scan data is sent to the data system for display, storage, or both. The process of ramping the amplitude of the rf and dc voltages on the quadrupole rods in the mass analyzer to transmit ions from the lowest mass to the highest mass of a specified scan range.
selected ion monitoring (SIM)
A scan type where the mass spectrometer acquires and records ion current following the isolation of a list of singular mass-to-charge ratio values.
selected reaction monitoring (SRM)
A scan type with two stages of mass analysis and in which a particular reaction or set of reactions, such as the fragmentation of an ion or the loss of a neutral moiety, is monitored. In SRM a limited number of product ions is monitored.
sheath gas
The inner coaxial gas (nitrogen), which is used in the API source to help nebulize the sample solution into a fine mist as the sample solution exits the ESI, H-ESI or APCI nozzle.
signal-to-noise ratio (S/N)
The ratio of the signal height (S) to the noise height (N). The signal height is the baseline corrected peak height. The noise height is the peak-to-peak height of the baseline noise.
sweep gas
Nitrogen gas that flows out from the gap between the sweep cone and the ion transfer tube into the API source. Sweep gas aids in solvent declustering and adduct reduction.
syringe pump
A device that delivers a solution from a syringe at a specified rate.
turbomolecular pump (TMP)
A vacuum pump that provides a high vacuum for the mass spectrometer and detector system.
vacuum manifold
A thick-walled, aluminum chamber with machined flanges on the front and sides and various electrical feedthroughs and gas inlets that encloses the API stack, ion optics, mass analyzer, and ion detection system.
vacuum system
Components associated with lowering the pressure in the mass spectrometer. A vacuum system includes the vacuum manifold, pumps, pressure gauges, and associated electronics.