The TSQ Series II mass spectrometers operate in a variety of scan types. The most common can be divided into two categories—single mass spectrometry (MS) scan types and MS/MS scan types. The scan types in each category are as follows:
- MS scan types: Full scan Q1, Full scan Q3, SIM Q1, and SIM Q3
- MS/MS scan types: Product ion scan, Precursor ion scan, Neutral loss scan, Selected reaction monitoring (SRM), and Quantitation enhanced data-dependent (QED)
See these topics: