You can control the divert/inject valve by using the mass spectrometry data system or the control buttons on the valve:
- Use the data system to specify the parameters in the Divert Valve Properties pane of the Method Editor for the MS. For instructions, refer to the Method Editor Help for the MS.
- Use the valve’s control buttons to divert the LC flow between the MS and waste when the valve is in the divert valve configuration, or switch between load and inject modes when the valve is in the loop injector configuration. For instructions, refer to the manufacturer’s manual.
| No. | Description | No. | Description |
|---|---|---|---|
1 | Valve position indicator | 3 | Valve control buttons |
2 | 6-port, 2-position valve |