The following figure shows the internal flow paths for the two valve positions.
  • In the first position, port 1 connects internally to port 2, port 3 connects to port 4, and port 5 connects to port 6.
  • In the second position, the valve rotates clockwise one position so that port 1 connects internally to port 6, port 2 connects to port 3, and port 4 connects to port 5.
NOTE
The Method Editor application identifies the valve’s two positions as 1–2 (port 1 to 2) and 1–6 (port 1 to 6).
Divert/Inject valve positions
12
No.DescriptionNo.Description
1
Position 1–2: Internal connection path in light gray
2
Position 1–6: The rotor switches between position 1–2 to 1–6