You can manually save the current settings for the source settings and scan parameters in the Favorites pane.
- To create a favorite state
- To load settings only or apply a favorite state
- To rename a favorite state
- To delete a favorite state
To create a favorite state
Procedure
- In the Tune window, modify the parameters in the Ion Source or Define Scan pane.
- Select Apply or Export.
- Select the Favorites tab to display the Favorites pane.
Figure Favorites pane - Select Save Current State, and then type a unique name in the box.
Figure State name box - Select Save Current State again to save the state.
- The new favorite state appears first in the Favorites list. You can enter up to 100 states.
To load settings only or apply a favorite state
Procedure
- Under User Settings, right-click the state name, and then choose one of the following:
- Load—to only display the key parameters in the applicable parameter boxes.
- Apply—to submit the key parameters to the MS.
To rename a favorite state
Procedure
- Under User Settings, right-click the state name, and then select Rename.
- Type a different name and press ENTER.
To delete a favorite state
Procedure
- Under User Settings, right-click the state name, and then select Delete.