To add a change record to the History pane, select Apply in the Ion Source or the Define Scan pane.
The Tune application adds a change record to the History pane. The change record records all changes to the instrument state that originated from the Tune application.
Change records in the History pane work as follows:
  • The Tune application creates a change record when you change parameters in the Ion Source or Define Scan pane and then select Apply.
  • The History pane displays the change records as sub-items under the date when they were created. The maximum number of change records is 100.
  • You can display a change record’s parameters by either double-clicking the record or right-clicking it and choosing Load. Parameters that are in bold differ from their currently set values.
  • You can submit a change record’s parameters to the MS by either double-clicking the record or by right-clicking it and choosing Apply.
  • A change record is inactive if the source type of the change record differs from the current source type.