The following table the lists the various system readback states that are located on the top right portion of the Tune application window.
Icon | Background color | Meaning |
---|---|---|
Green | The system parameters are within tolerance. | |
Green | The system is initializing. | |
Amber | One or more settings are changing. | |
Red | An error has occurred or the mass calibration has expired. | |
Gray | The MS is off. | |
Gray | There is no communication between the MS and the data system. |