The following table shows the options for the system control buttons available at the top of the Tune application window.
Button | Function | Description |
---|---|---|
Power mode | Sets the power mode for the MS (On, Standby, or Off). The center of the selected icon changes from white to green. See Set the instrument power mode. | |
Ion polarity mode | Sets the ion polarity mode to either Positive or Negative. | |
Data type | Sets the data type to either Centroid or Profile mode. | |
Spectrum scan averaging | To turn On or Off the scan averaging feature. If scan averaging is turned On, set the scan averaging value. | |
Divert/inject valve | Sets the valve module and valve position of the divert/injection valve. | |
Syringe pump | Select Syringe On (Off ) to switch between on and off. To set the syringe pump parameters Procedure
|