Procedure
- From the Windows™ taskbar, choose Start
. Then select Thermo Instruments > TSQ > Model Tune.
For information about the buttons and icons in the Tune application and what they control, refer to the Tune Help.
No. | Description | No. | Description |
|---|---|---|---|
1 | Three power mode icons (on/standby/off ) | 6 | Controls for graphs |
2 | Manual data acquisition pane | 7 | Plot Chromatogram tool |
3 | Instrument readback status | 8 | Define Scan pane |
4 | Set Options or View Help icon | 9 | Diagnostics icon |
5 | Status, History, and Favorites panes |