CAUTION
Avoid exposure to potentially harmful materials
By law, producers and suppliers of chemical compounds are required to provide their customers with the most current health and safety information in the form of Material Safety Data Sheets (MSDSs) or Safety Data Sheet (SDS). The MSDSs and SDSs must be freely available to lab personnel to examine at any time. These data sheets describe the chemicals and summarize information on the hazard and toxicity of specific chemical compounds. They also provide information on the proper handling of compounds, first aid for accidental exposure, and procedures to remedy spills or leaks.
CAUTION
Read the MSDS or SDS for each chemical you use. Store and handle all chemicals in accordance with standard safety procedures. Always wear protective gloves and safety glasses when you use solvents or corrosives. Also, contain waste streams, use proper ventilation, and dispose of all laboratory reagents according to the directions in the MSDS or SDS.
The MS requires few tools to perform routine maintenance procedures. You can remove and disassemble many of the components by hand.
The TSQ Source Installation Kit includes the custom tools that you use to remove and install the ion transfer tube and L0 lens.
The following table lists the necessary chemicals, tools, and equipment for maintaining the instrument.
Description | Part number |
|---|---|
Chemicals | |
Liquinox™ detergent |
|
Methanol, UHPLC/MS grade | |
Nitrogen gas, clean and dry | — |
Water, UHPLC/MS grade | |
Tools | |
Ion transfer tube removal tool In these models, the ion transfer tube is secured with a quarter-turn pin behind its nose cone. Note: This tool is provided in the TSQ Source Installation kit. | 70005-20972 |
Ion transfer tube removal tool In these models the ion transfer tube is secured with a threaded nose cone. Note: This tool is provided in the TSQ Source Installation kit. | 70111-20258 |
L0 lens removal tool | 70005-20900 |
Screwdriver, Phillips #2 (M3) | — |
(Optional) Toothbrush, soft (or similar tool) | — |
(Optional) Tweezers, plastic (or similar tool) | — |
Equipment | |
Beaker or graduated cylinder (for use with methanol) | — |
Chamois-tipped swabs | 00725-01-00028 |
Gloves, lint-free and powder-free |
|
Industrial tissues, lint-free | — |
Magnification device | — |
MICRO-MESH™ polishing swabs, 6000 grit (light purple color), 2.25 in. long | — |
Sonicator | — |