Performing routine maintenance procedures ensures optimum performance of the MS system.
The following table lists the maintenance procedures and their recommended frequency.
MS component | Procedure | Recommended frequency | Reference topic |
---|---|---|---|
API source (external) | Flush to clean the sample transfer line, sample tube, and spray insert. | After acquiring samples | |
Clean the API source housing. | As needed | ||
Clean the APPI fan filter. | As needed | Refer to the ion source guide | |
Replace the APPI lamp. | As needed | ||
Replace the H-ESI needle insert. | When the metal needle is obstructed | ||
Replace the APCI fused-silica tubing. | When the tubing is obstructed | ||
API source interface | Clean the ion sweep cone and the spray cone. | Weekly or more often depending on the analytical conditions | |
Remove and clean the ion transfer tube. | Weekly or when the ion transfer tube bore becomes contaminated or obstructed | ||
Replace the ion transfer tube. | If the bore becomes corroded or blocked | ||
Clean the exit lens or the RF lens. | As needed, depending on analytical conditions | ||
Clean the MP00 multipole and L0 lens. | As needed, depending on the analytical conditions | ||
Forepump (each) | Purge (ballast) the oil of volatile solvents and check for leaks. | Weekly | Manufacturer’s manual |
Add oil. | As needed, based on the oil level | ||
Change the oil. | Every 12 months of typical use, or when the oil becomes cloudy or discolored | ||
Cooling fans | Clean the air filter. | Every 4 months |