After you have removed the components of the API source interface cage for cleaning, follow these topics as applicable for the reinstallation:
- Reassemble the MP00 assembly
- Reinstall the lenses and the MP00 assembly
- Reinstall the lenses and the MP00 assembly (TSQ Fortis Plus and TSQ Fortis only)
Reassemble the MP00 assembly
Follow this procedure to reassemble the MP00 Assembly.
Procedure
- Align the MP00 multipole with the alignment pin in the mounting cage, and then insert the multipole into the cage.
Inserting the MP00 multipole into the mounting cage No. Description No. Description 1
Alignment pin
2
MP00 multipole
Reinstall the lenses and the MP00 assembly
NOTE
This procedure applies to all TSQ Series II instruments excluding the TSQ Fortis Plus and TSQ Fortis models.
Follow this procedure to reinstall the RF lens, the exit lens, the MP00 assembly, and L0 lens in the source interface cage.
Procedure
- Reassemble the MP00 assembly.
- Attach the exit lens to the RF lens, and then reinsert the RF lens into the cage. See Remove the MP00 assembly, the RF lens, and the exit lens from the source interface cage for reference, if needed.
- Attach the MP00 assembly to the cage, and then attach the L0 lens.
Reinstall the lenses and the MP00 assembly (TSQ Fortis Plus and TSQ Fortis only)
NOTE
This procedure applies only to the TSQ Fortis Plus and TSQ Fortis instruments.
Follow this procedure to reinstall the lenses and MP00 assembly.
Required tool |
---|
L0 lens removal tool |
Procedure
- Reassemble the MP00 assembly. See Reinstall the components of the API source interface cage.
- Align the two slots in the outer ring of the L0 lens with the tabs on the mounting cage. Then, use the L0 lens removal tool to turn the L0 lens by a quarter turn.
- Align the tube lens pin to the port in the inner circle of the API source interface cage, and then insert the lens into the cage and snap it into place.
API source interface cage with the lenses removed No. Description No. Description 1
Mounting port for the skimmer
2
Mounting port for the tube lens
Installing the tube lens No. Description No. Description 1
Tube lens pin
2
Mounting port for the tube lens
- Align the skimmer pin to the mounting port in the cage, and then insert the skimmer into the cage.
Inserting the skimmer into the cage No. Description No. Description 1
Skimmer pin
2
Mounting port for the skimmer
- Reconnect the MP00 assembly to the cage. Use a 2 mm Allen wrench to tighten the two screws that secure the assembly to the cage.