This procedure applies to the TSQ Fortis Plus and TSQ Fortis MS's only.
Preconditions
The MS is vented and cooled to room temperature.
Required tools
  • 2 mm Allen wrench
  • L0 lens removal tool
  • Small flat-head screwdriver
Procedure
  1. Wearing clean, lint-free and powder-free gloves, remove the source interface cage from the MS.
  2. API source interface (TSQ Fortis Plus and TSQ Fortis)
    Figure API source interface (TSQ Fortis Plus and TSQ Fortis)
  3. Remove the two Allen screws that secure the MP00 assembly to the cage using a 2 mm Allen wrench.
  4. Removing the Allen screws that secure the MP00 assembly to the cage
    Figure Removing the Allen screws that secure the MP00 assembly to the cage
  5. Pull the MP00 assembly off of the cage and place it on a clean and lint-free surface.
  6. MP00 assembly removed from the cage (TSQ Fortis Plus and TSQ Fortis)
    Figure MP00 assembly removed from the cage (TSQ Fortis Plus and TSQ Fortis)
  7. Use a small flat-head screwdriver to detach the skimmer from the cage. Then lift the skimmer evenly from both sides to prevent bending the voltage contact pin and remove it from the cage.
  8. CAUTION
    Use extra care when removing the skimmer from the cage because the voltage contact pin can easily bend.
  9. Removing the skimmer from the cage (TSQ Fortis Plus and TSQ Fortis)
    Figure Removing the skimmer from the cage (TSQ Fortis Plus and TSQ Fortis)
  10. Figure Skimmer removed from the cage (TSQ Fortis Plus and TSQ Fortis)
    123
    No.DescriptionNo.Description
    1
    Skimmer
    3
    Tube lens
    2
    Voltage contact pin
  11. Remove the tube lens by pushing it out of the cage.
  12. NOTE
    It can take quite a bit of force to push out the tube lens the first few times you remove it.
  13. Pushing the tube lens out of the cage (TSQ Fortis Plus and TSQ Fortis)
    Figure Pushing the tube lens out of the cage (TSQ Fortis Plus and TSQ Fortis)
  14. To remove the L0 lens from the MP00 assembly, use the L0 lens removal tool or plastic tweezers. Rotate the lens by a quarter turn to release it from the two tabs.
  15. Figure Using the L0 lens removal tool to rotate L0 lens
    123
    No.DescriptionNo.Description
    1
    L0 lens secured to the MP00 assembly by two tabs
    3
    L0 lens released (but not removed) from the MP00 assembly
    2
    Rotating the L0 lens by a quarter turn
  16. Push the MP00 multipole out of the mounting cage.
  17. Pushing the MP00 multipole out of the mounting cage
    Figure Pushing the MP00 multipole out of the mounting cage
  18. Place the components on a clean and lint-free surface.
  19. Tube lens, skimmer, L0 lens, and MP00 multipole (from left-to-right)
    Figure Tube lens, skimmer, L0 lens, and MP00 multipole (from left-to-right)