Procedure
- Remove the sweep cone and the ion transfer tube. See Remove the ion sweep cone and Remove the ion transfer tube.
- Soak a lint-free tissue or a chamois-tipped swab in a 50:50 solution of methanol/water, and then clean the exterior surface of the spray cone.
Figure Spray cone, seal, ion transfer tube, and ion sweep cone No. Description No. Description 1
API cone seal
4
Ion transfer tube
2
Spray cone
5
Gas inlet on the ion sweep cone
3
Graphite Vespel seal
- If you see a leak at the seal, do the following:
- Remove and inspect the seal located in the spray cone under the entrance end of the ion transfer tube.
- Clean the seal with a wipe with methanol or replace the seal, if necessary.
- Using a magnification device, inspect the components for any residual lint or particulates.
NOTE
Inspect the inside surfaces and edges for the presence of lint or particulates. If present, use plastic tweezers or a similar tool to remove them.
- Reinstall the seal in the spray cone.