Removing or installing the ion transfer tube requires the custom tool that is provided in the TSQ Source Installation Kit. This kit includes two ion transfer tube removal tools. The tool you need depends on whether the ion transfer tube is secured by a quarter-turn pin or a threaded fitting.
Model | Attachment type |
|---|---|
TSQ Altis Plus, TSQ Altis, and TSQ Quantiva | Quarter-turn pin |
TSQ Quantis Plus, TSQ Quantis, TSQ Fortis Plus, | Threaded fitting |
Follow the appropriate procedure for your TSQ Series II MS:
- Remove an ion transfer tube that is secured with a quarter-turn pin
- Remove an ion transfer tube that is secured with a threaded fitting
Remove an ion transfer tube that is secured with a quarter-turn pin
(TSQ Altis Plus, TSQ Altis, and TSQ Quantiva)
Procedure
- Remove the API source housing. See Maintain the API source housing.
- Remove the ion sweep cone by grasping its outer ridges and pulling it away from API source interface. See Remove the ion sweep cone.
- Align the flat end of the 1/4 turn ion transfer tube removal tool with the flat edges on the ion transfer tube’s nose cone.
- Rotate the tube counterclockwise by a quarter turn to release the pins behind the nose cone that secure the tube to the spray cone.
Figure Turning the nose cone counterclockwise by a quarter turn - Use the other end of the tool to pull the tube out of the API source interface.
Figure Pulling the transfer tube out of the API source interface No. Description No. Description 1
Use this end of the tool to rotate the tube counterclockwise by a 1/4 turn, releasing it from the spray cone.
2
Use this end of the tool to hook onto the released tube and pull it out of the spray cone.
To clean the ion transfer tube, go to Clean the ion sweep cone and the ion transfer tube.
Remove an ion transfer tube that is secured with a threaded fitting
(TSQ Quantis Plus, TSQ Quantis, TSQ Fortis Plus, TSQ Fortis, and TSQ Endura)
Procedure
- Align the hook end of the ion transfer tube removal tool with the flat edges of the ion transfer tube’s nose cone.
- Rotate the ion transfer tube counterclockwise until its threaded nose cone is free of the API interface.
TIP
If necessary, insert a hex key through the side hole in the tool and use it for leverage.
Figure Ion transfer tube removal tool (for tube with a threaded nose cone) No. Description No. Description 1
API source interface
3
Side hole for leveraging tool
2
Hook end of the tool aligned to the flat edges of the ion transfer tube’s nose
- Hook the tool onto the back of the ion transfer tube’s nose cone, and then pull the tube out of the spray cone.
Figure Pulling the tube out of the spray cone