Because buffer salts and high concentration samples can cause blockages, you must clean the ion sweep cone, the ion transfer tube, and the spray cone on a regular basis.
If the pressure in the ion transfer tube and RF lens region (as measured by the Source Pressure gauge) drops below the pressure listed in the following table, the ion transfer tube is probably blocked, and you might need to replace it.
Model | Pressure reading |
---|---|
TSQ Altis Plus , TSQ Altis, and TSQ Quantiva | <2 Torr |
All other models in the TSQ Series II product line | <1 Torr |
Clean the following external components of the API source interface at the same time:
- Ion sweep cone
- External surface of the spray cone
- Ion transfer tube
Replace the ion transfer tube if the cleaning procedure does not remove the blockage.
CAUTION
To prevent corrosion, do not use nitric acid to clean metal parts.
TIP
You do not have to vent the system to remove the ion sweep cone or ion transfer tube. You do not need to remove the spray cone to clean its external surface.
To maintain the ion sweep cone, the ion transfer tube, and the spray cone, follow these steps: