The LC flow rate range for nanospray ionization is 10 to 1,000 nL/min.
Spray voltage (V) | Sweep gas (arb) | Ion transfer tube temp (°C) |
|---|---|---|
Positive mode: 1,200 | 0 | 325 |
Negative mode: 600 | 0 | 325 |
The LC flow rate range for nanospray ionization is 10 to 1,000 nL/min.
Spray voltage (V) | Sweep gas (arb) | Ion transfer tube temp (°C) |
|---|---|---|
Positive mode: 1,200 | 0 | 325 |
Negative mode: 600 | 0 | 325 |