For the APCI ion source, the recommended setting for the Corona Discharge Voltage depends on the polarity:
  • Positive ion discharge current: 4 μA
  • Negative ion discharge current: 10 μA
Table Recommended LC/APCI/MS parameters
LC Flow Rates (μL/min)
<=14.99
15–99.99
100–199.99
200–499.99
500–999.99
1,000–3,000
Sheath gas (arb)
10
15
25
45
45
45
Aux gas (arb)
2
5
5
5
10
10
Sweep gas (arb)
0
0
0
1
2
2
Vaporizer temp (°C)
0
275
325
350
400
400
Nitrogen consumption (L/min)
4
7
8
12
16
16
Ion transfer tube temp (°C)
TSQ Altis Plus,
TSQ Altis,
and TSQ Quantiva
300
300
300
325
375
375
TSQ Quantis Plus,
TSQ Quantis,
and TSQ Endura
275
275
275
300
325
350
Fortis Plus and Fortis
250
250
250
275
300
325