The mass spectra of the EMRS in positive and negative ion polarity modes have peaks at m/z values close to the theoretical values in the following tables.
IMPORTANT
If you observe interfering peaks in the EMRS spectrum that are within ±10 Da of any of these calibration masses, follow the procedure Flush the inlet components. Ensure that the interfering masses show less than 25 percent of the intensity of the calibrant ions.
Component | Positive mode (m/z) |
|---|---|
Imidazole | 69.04 |
Triethylamine | 102.13 |
1,8-Bis(diethylamino)napthalene | 215.15 |
Hexakis(2,2-difluoroethoxy)phosphazene | 622.03 |
Hexakis(2,2,3,3-tetrafluoropropoxy)phosphazene | 922.01 |
Hexakis(1h,1h,5h-octafluoropentoxy)phosphazene | 1521.97 |
Component | Positive mode (m/z) |
|---|---|
Fragment of TFA | 69.00 |
Trifluoroacetic acid (TFA) | 112.991) |
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine | 302.002) |
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine | 601.982) |
Hexakis(2,2,3,3-tetrafluoropropoxy)phosphazene | 1033.993) |
Hexakis(1h,1h,5h-octafluoropentoxy)phosphazene | 1633.953) |